Assay Details
| Gene Symbol: ISY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252375.1 | NP_001239304.1 | 130 | 113 | ||
| XM_005632027.3 | XP_005632084.1 | 531 | 113 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ISY1
Genomic Map