Assay Details
| Gene Symbol: PGM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117685.2 | XP_013973160.2 | 5 - 6 | 674 | 84 | |
| XM_022421511.1 | XP_022277219.1 | 5 - 6 | 650 | 84 | ||
| XM_022421512.1 | XP_022277220.1 | 5 - 6 | 647 | 84 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PGM5
Genomic Map