Assay Details
| Gene Symbol: SELENOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114757.1 | NP_001108229.1 | 2 | 124 | 96 | |
| GenBank mRNA | DN370248.1 | - | 2 | 121 | 96 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SELENOS
Genomic Map