Assay Details
Gene Symbol: TSPAN17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619177.3 | XP_005619234.1 | 2 - 3 | 302 | 154 | |
XM_005619179.2 | XP_005619236.1 | 2 - 3 | 95 | 154 | ||
XM_022417675.1 | XP_022273383.1 | 2 - 3 | 302 | 154 | ||
XM_022417676.1 | XP_022273384.1 | 2 - 3 | 303 | 154 | ||
XM_860797.5 | XP_865890.1 | 2 - 3 | 302 | 154 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
