Assay Details
| Gene Symbol: CDC42SE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039043.1 | NP_001034132.1 | 3 - 4 | 485 | 154 | |
| XM_014120563.2 | XP_013976038.1 | 3 - 4 | 556 | 154 | ||
| GenBank mRNA | DN267318.1 | - | 3 - 4 | 485 | 154 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CDC42SE1
Genomic Map