Assay Details
Gene Symbol: LGALS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284486.1 | NP_001271415.1 | 798 | 184 | ||
XM_005618787.2 | XP_005618844.1 | 1724 | 184 | |||
XM_005618788.3 | XP_005618845.1 | 1523 | 184 | |||
XM_005618790.3 | XP_005618847.1 | 1425 | 184 | |||
XM_005618791.3 | XP_005618848.1 | 1084 | 184 | |||
XM_005618792.3 | XP_005618849.1 | 1086 | 184 | |||
XM_014112703.2 | XP_013968178.1 | 1559 | 184 | |||
XM_014112704.2 | XP_013968179.1 | 1469 | 184 | |||
XM_014112705.2 | XP_013968180.1 | 1420 | 184 | |||
XM_022417336.1 | XP_022273044.1 | 1737 | 184 | |||
XM_022417337.1 | XP_022273045.1 | 1046 | 184 | |||
XM_022417338.1 | XP_022273046.1 | 1455 | 184 | |||
XM_536335.6 | XP_536335.2 | 1082 | 184 | |||
GenBank mRNA | HQ637389.1 | - | 798 | 184 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
