Assay Details
Gene Symbol: LUC7L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131056.1 | NP_001124528.1 | 737 | 164 | ||
XM_005629474.2 | XP_005629531.1 | 716 | 164 | |||
XM_005629475.3 | XP_005629532.1 | 483 | 164 | |||
XM_005629477.3 | XP_005629534.1 | 1060 | 164 | |||
XM_005629478.3 | XP_005629535.1 | 328 | 164 | |||
XM_014119776.2 | XP_013975251.1 | 491 | 164 | |||
XM_014119777.2 | XP_013975252.1 | 522 | 164 | |||
XM_022403592.1 | XP_022259300.1 | 1219 | 164 | |||
XM_022403593.1 | XP_022259301.1 | 764 | 164 | |||
XM_022403594.1 | XP_022259302.1 | 1128 | 164 | |||
XM_022403595.1 | XP_022259303.1 | 790 | 164 | |||
XM_022403596.1 | XP_022259304.1 | 515 | 164 | |||
XM_022403597.1 | XP_022259305.1 | 393 | 164 | |||
XM_022403598.1 | XP_022259306.1 | 617 | 164 | |||
XM_022403599.1 | XP_022259307.1 | 462 | 164 | |||
XM_022403600.1 | XP_022259308.1 | 385 | 164 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
