Assay Details
Gene Symbol: KIAA0513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620637.3 | XP_005620694.1 | 7 | 1024 | 159 | |
XM_005620638.3 | XP_005620695.1 | 6 | 1003 | 159 | ||
XM_014113985.2 | XP_013969460.1 | 7 | 1024 | 159 | ||
XM_022419108.1 | XP_022274816.1 | 7 | 1024 | 159 | ||
XM_546795.6 | XP_546795.3 | 6 | 933 | 159 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
