Assay Details
Gene Symbol: ADAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624829.3 | XP_005624886.1 | 9 - 10 | 1011 | 190 | |
XM_014116760.2 | XP_013972235.1 | 9 - 10 | 1011 | 190 | ||
XM_014116762.2 | XP_013972237.1 | 8 - 9 | 927 | 190 | ||
XM_014116763.2 | XP_013972238.1 | 8 - 9 | 864 | 190 | ||
XM_022423318.1 | XP_022279026.1 | 9 - 10 | 913 | 190 | ||
XM_022423319.1 | XP_022279027.1 | 8 - 9 | 772 | 190 | ||
XM_848333.5 | XP_853426.2 | 9 - 10 | 1011 | 190 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
