Assay Details
| Gene Symbol: TNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014113528.2 | XP_013969003.1 | 5 - 6 | 809 | 155 | |
| XM_014113529.2 | XP_013969004.1 | 5 - 6 | 809 | 155 | ||
| XM_022418510.1 | XP_022274218.1 | 5 - 6 | 718 | 155 | ||
| XM_022418511.1 | XP_022274219.1 | 5 - 6 | 809 | 155 | ||
More Information
Set Membership: |
|

TNK1
Genomic Map