Assay Details
Gene Symbol: ST6GALNAC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625323.3 | XP_005625380.2 | 2 - 3 | 306 | 93 | |
XM_005625328.3 | XP_005625385.1 | 3 - 4 | 279 | 93 | ||
XM_014117036.2 | XP_013972511.1 | 2 - 3 | 323 | 93 | ||
XM_022423778.1 | XP_022279486.1 | 2 - 3 | 308 | 93 | ||
XM_022423779.1 | XP_022279487.1 | 3 - 4 | 415 | 93 | ||
XM_022423780.1 | XP_022279488.1 | 2 - 3 | 561 | 93 | ||
XM_022423782.1 | XP_022279490.1 | 3 - 4 | 407 | 93 | ||
XM_022423783.1 | XP_022279491.1 | 2 - 3 | 226 | 93 | ||
XM_022423784.1 | XP_022279492.1 | 2 - 3 | 328 | 93 | ||
XM_022423785.1 | XP_022279493.1 | 2 - 3 | 235 | 93 | ||
XM_022423787.1 | XP_022279495.1 | 2 - 3 | 338 | 93 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_005625323.3, XM_014117036.2, XM_022423778.1, XM_022423782.1, XM_022423783.1, XM_022423784.1, XM_022423785.1, XM_022423787.1 transcripts.
This assay is in the 5'UTR region of the XM_005625323.3, XM_014117036.2, XM_022423778.1, XM_022423782.1, XM_022423783.1, XM_022423784.1, XM_022423785.1, XM_022423787.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
