Assay Details
| Gene Symbol: SYT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427463.1 | XP_022283171.1 | 4 - 5 | 762 | 151 | |
| XM_022427470.1 | XP_022283178.1 | 5 - 6 | 708 | 151 | ||
| XM_533585.6 | XP_533585.1 | 5 - 6 | 724 | 151 | ||
More Information
Set Membership: |
|

SYT5
Genomic Map