Assay Details
| Gene Symbol: CIART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639643.4 | XP_003639691.1 | 5 - 6 | 1202 | 165 | |
| XM_005630746.3 | XP_005630803.1 | 5 - 6 | 799 | 165 | ||
More Information
Set Membership: |
|

CIART
Genomic Map