Assay Details
| Gene Symbol: PEG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172543.2 | NP_001166014.2 | 2 | 1042 | 179 | |
| NM_001172544.2 | NP_001166015.2 | 2 | 1042 | 179 | ||
More Information
Set Membership: |
|

PEG10
Genomic Map