Assay Details
| Gene Symbol: PLEKHA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167698.1 | NP_001161170.1 | 10 - 11 | 1407 | 161 | |
| XM_022427142.1 | XP_022282850.1 | 10 - 11 | 1207 | 161 | ||
| XM_022427143.1 | XP_022282851.1 | 10 - 11 | 1075 | 161 | ||
| XM_022427144.1 | XP_022282852.1 | 10 - 11 | 1221 | 161 | ||
| XM_022427145.1 | XP_022282853.1 | 10 - 11 | 1203 | 161 | ||
| GenBank mRNA | GU204956.1 | - | 10 - 11 | 1093 | 161 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PLEKHA8
Genomic Map