Assay Details
Gene Symbol: CCNYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640591.3 | XP_005640648.1 | 7 - 8 | 702 | 145 | |
XM_545613.6 | XP_545613.3 | 5 - 6 | 554 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
