Assay Details
| Gene Symbol: TMEM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639069.4 | XP_003639117.1 | 2 | 353 | 146 | |
| XM_005620692.3 | XP_005620749.1 | 2 | 353 | 215 | ||
| XM_014114004.2 | XP_013969479.1 | 2 | 367 | 215 | ||
| XM_014114005.2 | XP_013969480.1 | 2 | 612 | 215 | ||
| XM_022419182.1 | XP_022274890.1 | 2 | 367 | 146 | ||
| XM_022419183.1 | XP_022274891.1 | 2 | 612 | 146 | ||
More Information
Set Membership: |
|

TMEM170A
Genomic Map