Assay Details
| Gene Symbol: NIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111227.2 | XP_013966702.1 | 6 - 7 | 740 | 163 | |
| XM_022415431.1 | XP_022271139.1 | 6 - 7 | 847 | 163 | ||
| XM_022415432.1 | XP_022271140.1 | 6 - 7 | 857 | 163 | ||
| XM_022415433.1 | XP_022271141.1 | 6 - 7 | 740 | 163 | ||
| XM_022415434.1 | XP_022271142.1 | 6 - 7 | 884 | 163 | ||
| XM_536134.6 | XP_536134.3 | 6 - 7 | 820 | 163 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NIT1
Genomic Map