Assay Details
| Gene Symbol: HYAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633189.3 | XP_005633246.2 | 2 | 387 | 149 | |
| XM_014122054.2 | XP_013977529.1 | 2 | 387 | 149 | ||
More Information
Set Membership: |
|

HYAL3
Genomic Map