Assay Details
Gene Symbol: SLC35A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003059.2 | NP_001003059.2 | 3 | 350 | 145 | |
XM_005640971.3 | XP_005641028.1 | 3 | 669 | 145 | ||
XM_022415485.1 | XP_022271193.1 | 3 | 789 | 145 | ||
XM_022415486.1 | XP_022271194.1 | 2 | 488 | 145 | ||
XM_022415487.1 | XP_022271195.1 | 2 | 488 | 145 | ||
GenBank mRNA | AM110117.1 | - | 2 | 167 | 145 | |
AY064406.2 | - | 3 | 350 | 145 | ||
FN825775.1 | - | 3 | 353 | 145 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
