Assay Details
| Gene Symbol: UROD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022428022.1 | XP_022283730.1 | 8 - 9 | 924 | 107 | |
| XM_532602.6 | XP_532602.3 | 9 - 10 | 1055 | 107 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UROD
Genomic Map