Assay Details
| Gene Symbol: ADORA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002944.1 | NP_001002944.1 | 2 | 526 | 131 | |
| GenBank mRNA | AY313204.1 | - | 2 | 526 | 131 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ADORA2B
Genomic Map