Assay Details
Gene Symbol: LOC477556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022410501.1 | XP_022266209.1 | 2 - 3 | 346 | 173 | |
XM_534751.6 | XP_534751.3 | 3 - 4 | 435 | 173 | ||
GenBank mRNA | KM233847.1 | - | 3 - 4 | 393 | 173 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_022410501.1 transcripts.
This assay is in the 5'UTR region of the XM_022410501.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
