Assay Details
Gene Symbol: PTER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617099.3 | XP_005617156.1 | 636 | 148 | ||
XM_005617101.2 | XP_005617158.1 | 548 | 148 | |||
XM_005617102.3 | XP_005617159.1 | 548 | 148 | |||
XM_843558.5 | XP_848651.1 | 548 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
