Assay Details
| Gene Symbol: CXCR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048026.1 | NP_001041491.1 | 2 | 229 | 116 | |
| GenBank mRNA | DQ182699.1 | - | 2 | 229 | 116 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CXCR4
Genomic Map