Assay Details
| Gene Symbol: FGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629368.3 | XP_005629425.1 | 7 - 8 | 1014 | 121 | |
| XM_532698.6 | XP_532698.2 | 7 - 8 | 1012 | 121 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FGG
Genomic Map