Assay Details
Gene Symbol: DCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111894.2 | XP_013967369.1 | 2 - 3 | 618 | 135 | |
XM_014111895.2 | XP_013967370.1 | 2 - 3 | 535 | 135 | ||
XM_022415817.1 | XP_022271525.1 | 2 - 3 | 619 | 135 | ||
XM_022415818.1 | XP_022271526.1 | 2 - 3 | 619 | 135 | ||
XM_022415819.1 | XP_022271527.1 | 2 - 3 | 618 | 135 | ||
XM_022415820.1 | XP_022271528.1 | 2 - 3 | 618 | 135 | ||
XM_022415821.1 | XP_022271529.1 | 2 - 3 | 619 | 135 | ||
XM_022415822.1 | XP_022271530.1 | 2 - 3 | 620 | 135 | ||
XM_022415823.1 | XP_022271531.1 | 2 - 3 | 619 | 135 | ||
XM_022415824.1 | XP_022271532.1 | 2 - 3 | 620 | 135 | ||
XM_022415825.1 | XP_022271533.1 | 3 - 4 | 651 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
