Assay Details
| Gene Symbol: IGF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195403.1 | NP_001182332.1 | 3 - 4 | 462 | 135 | |
| NM_001195825.1 | NP_001182754.1 | 3 - 4 | 471 | 135 | ||
| NM_001195826.1 | NP_001182755.1 | 3 - 4 | 421 | 135 | ||
| NM_001195827.1 | NP_001182756.1 | 3 - 4 | 412 | 135 | ||
| XM_005631053.3 | XP_005631110.1 | 4 - 5 | 560 | 135 | ||
| XM_014120867.1 | XP_013976342.1 | 3 - 4 | 931 | 135 | ||
| XM_014120868.2 | XP_013976343.1 | 4 - 5 | 435 | 135 | ||
| XM_022405072.1 | XP_022260780.1 | 4 - 5 | 569 | 135 | ||
| XM_022405074.1 | XP_022260782.1 | 3 - 4 | 423 | 135 | ||
| XM_022405075.1 | XP_022260783.1 | 3 - 4 | 405 | 135 | ||
| GenBank mRNA | FJ159123.1 | - | 3 - 4 | 390 | 135 | |
| FJ159124.1 | - | 3 - 4 | 408 | 135 | ||
| FJ159125.1 | - | 3 - 4 | 399 | 135 | ||
| FJ159126.1 | - | 3 - 4 | 417 | 135 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IGF2
Genomic Map