Assay Details
| Gene Symbol: TUT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427898.1 | XP_022283606.1 | 26 - 27 | 4413 | 65 | |
| XM_022427899.1 | XP_022283607.1 | 26 - 27 | 4413 | 65 | ||
| XM_022427900.1 | XP_022283608.1 | 26 - 27 | 4410 | 65 | ||
| XM_022427901.1 | XP_022283609.1 | 26 - 27 | 4398 | 65 | ||
| XM_022427902.1 | XP_022283610.1 | 26 - 27 | 4413 | 65 | ||
| XM_022427903.1 | XP_022283611.1 | 26 - 27 | 4413 | 65 | ||
| XM_022427904.1 | XP_022283612.1 | 26 - 27 | 4410 | 65 | ||
| XM_022427905.1 | XP_022283613.1 | 26 - 27 | 4398 | 65 | ||
| XM_022427906.1 | XP_022283614.1 | 25 - 26 | 4326 | 65 | ||
| XM_022427907.1 | XP_022283615.1 | 25 - 26 | 4200 | 65 | ||
More Information
Set Membership: |
|

TUT4
Genomic Map