Assay Details
| Gene Symbol: NME2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024638.1 | NP_001019809.1 | 1 | 345 | 141 | |
| XM_005624342.3 | XP_005624399.1 | 553 | 141 | |||
| GenBank mRNA | AB207045.1 | - | 1 | 345 | 141 | |
| DN416219.1 | - | 1 | 385 | 141 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NME2
Genomic Map