Assay Details
Gene Symbol: SSTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031817.1 | NP_001026987.1 | 1044 | 97 | ||
XM_005623953.3 | XP_005624010.1 | 1320 | 97 | |||
XM_014116187.2 | XP_013971662.1 | 1473 | 97 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
