Assay Details
| Gene Symbol: ANXA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314118.1 | NP_001301047.1 | 6 - 7 | 497 | 103 | |
| XM_533303.6 | XP_533303.1 | 7 - 8 | 661 | 103 | ||
| GenBank mRNA | LC055191.1 | - | 6 - 7 | 497 | 103 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ANXA5
Genomic Map