Assay Details
Gene Symbol: CNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014122382.2 | XP_013977857.1 | 3 - 4 | 210 | 91 | |
XM_014122383.2 | XP_013977858.1 | 3 - 4 | 209 | 91 | ||
XM_542053.6 | XP_542053.1 | 3 - 4 | 352 | 91 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
