Assay Details
| Gene Symbol: ATP6V1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414013.1 | XP_022269721.1 | 7 - 8 | 1038 | 89 | |
| XM_545103.6 | XP_545103.1 | 7 - 8 | 1174 | 89 | ||
More Information
Set Membership: |
|

ATP6V1A
Genomic Map