Assay Details
| Gene Symbol: GPX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115135.1 | NP_001108607.1 | 1 - 2 | 237 | 158 | |
| GenBank mRNA | DN869773.1 | - | 1 - 2 | 700 | 158 | |
| DN872671.1 | - | 1 - 2 | 237 | 158 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GPX2
Genomic Map