Assay Details
Gene Symbol: ITPKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_547504.4 | XP_547504.2 | 2 - 3 | 2360 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
