Assay Details
| Gene Symbol: MYL6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117152.2 | XP_013972627.1 | 782 | 122 | ||
| XM_843490.5 | XP_848583.1 | 749 | 122 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MYL6B
Genomic Map