Assay Details
| Gene Symbol: FOXC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014113959.2 | XP_013969434.1 | 1392 | 149 | ||
| XM_022419103.1 | XP_022274811.1 | 1392 | 149 | |||
| XM_022419104.1 | XP_022274812.1 | 1392 | 149 | |||
More Information
Set Membership: |
|

FOXC2
Genomic Map