Assay Details
| Gene Symbol: DLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145071.1 | NP_001138543.1 | 2986 | 170 | ||
| NM_001313797.1 | NP_001300726.1 | 4514 | 170 | |||
| GenBank mRNA | FJ602870.1 | - | 2986 | 170 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DLC1
Genomic Map