Assay Details
Gene Symbol: PGRMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631808.2 | XP_005631865.2 | 808 | 164 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
