Assay Details
Gene Symbol: PMEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103216.1 | NP_001096686.1 | 470 | 134 | ||
XM_022423998.1 | XP_022279706.1 | 1567 | 134 | |||
XM_022423999.1 | XP_022279707.1 | 1156 | 134 | |||
XM_022424000.1 | XP_022279708.1 | 1329 | 134 | |||
XM_022424001.1 | XP_022279709.1 | 626 | 134 | |||
XM_022424002.1 | XP_022279710.1 | 1457 | 134 | |||
XM_022424003.1 | XP_022279711.1 | 1229 | 134 | |||
XM_022424004.1 | XP_022279712.1 | 837 | 134 | |||
XM_022424005.1 | XP_022279713.1 | 715 | 134 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
