Assay Details
| Gene Symbol: PLA2G15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002940.1 | NP_001002940.1 | 2 - 3 | 308 | 143 | |
| XM_005620606.3 | XP_005620663.1 | 2 - 3 | 1536 | 143 | ||
| GenBank mRNA | AY217754.1 | - | 2 - 3 | 308 | 143 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PLA2G15
Genomic Map