Assay Details
| Gene Symbol: PPP2R3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639829.4 | XP_003639877.1 | 6 - 7 | 3364 | 74 | |
| XM_005634462.3 | XP_005634519.1 | 6 - 7 | 3135 | 74 | ||
| XM_542792.7 | XP_542792.3 | 5 - 6 | 1014 | 74 | ||
More Information
Set Membership: |
|

PPP2R3A
Genomic Map