Assay Details
| Gene Symbol: PEX19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022415349.1 | XP_022271057.1 | 7 - 8 | 1053 | 105 | |
| XM_536130.6 | XP_536130.2 | 6 - 7 | 791 | 105 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PEX19
Genomic Map