Assay Details
| Gene Symbol: RPL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313778.1 | NP_001300707.1 | 3 - 4 | 369 | 156 | |
| XM_857783.4 | XP_862876.1 | 3 - 4 | 467 | 156 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RPL15
Genomic Map