Assay Details
| Gene Symbol: TIMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003082.1 | NP_001003082.1 | 471 | 136 | ||
| GenBank mRNA | AF112115.1 | - | 471 | 136 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TIMP2
Genomic Map