Assay Details
| Gene Symbol: HSP90B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003327.2 | NP_001003327.1 | 8 - 9 | 1189 | 115 | |
| GenBank mRNA | AB981782.1 | - | 8 - 9 | 1073 | 115 | |
| DN367458.1 | - | 2 - 3 | 246 | 115 | ||
| DN872944.1 | - | 2 - 3 | 145 | 115 | ||
| U01153.1 | - | 8 - 9 | 1192 | 115 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HSP90B1
Genomic Map