Assay Details
| Gene Symbol: SLC7A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284482.1 | NP_001271411.1 | 1234 | 158 | ||
| GenBank mRNA | AB636469.1 | - | 1234 | 158 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC7A5
Genomic Map