Assay Details
| Gene Symbol: DCAF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623570.3 | XP_005623627.1 | 8 - 9 | 959 | 68 | |
| XM_005623571.3 | XP_005623628.1 | 8 - 9 | 1087 | 68 | ||
| XM_547871.6 | XP_547871.3 | 8 - 9 | 1122 | 68 | ||
More Information
Set Membership: |
|

DCAF5
Genomic Map