Assay Details
| Gene Symbol: TPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197191.1 | NP_001184120.1 | 7 - 8 | 930 | 160 | |
| XM_005633302.3 | XP_005633359.1 | 8 - 9 | 1193 | 160 | ||
| GenBank mRNA | AB379810.1 | - | 7 - 8 | 930 | 160 | |
More Information
Set Membership: |
|

TPH1
Genomic Map